Learn More
Sino Biological ZIKV ZIKV-NS1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 40544MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-NS1 ( Catalog#40544-V07H; ALU33341.1; Val796-Leu1157). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1488