Learn More
Sino Biological Zika virus (ZIKV) (strain Zika SPH2015) E / Envelope protein (aa 291-696, His Tag)
Supplier:  Sino Biological 40543V08B5100
Encompass_Preferred
A DNA sequence encoding the Zika virus (strain Zika SPH2015) E (ALU33341.1, with mutation Thr 366 Ala, Gln 367 Gly, Trp 391 Arg, Leu 397 Arg) (Ile291-Thr696) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3704