Learn More
Sino Biological Zika virus (strain Zika SPH2015) E / Envelope protein (Domain I, His & MBP Tag)
Supplier:  Sino Biological 40543V10E100
Encompass_Preferred
A DNA sequence encoding the Zika virus (strain Zika SPH2015) E (ALU33341.1) (Ile291-Thr399 and Glu426-Gly485 and Ser576- Gly592) was expressed with a N-terminal polyhistidine tag followed by a MBP tag.
Catalog No. 50-255-3702