Learn More
Medchemexpress LLC Zileuton | 111406-87-2 | MFCD00866097 | 99.6% | C11H12N2O2S | 10MG
SDP
Catalog No. 5000258928
Encompass_Preferred
Zileuton is a purified small-molecule inhibitor of 5-lipoxygenase used as a research reagent to study leukotriene biology, inflammation, and asthma-related pathways. It is supplied as an analytical standard for in vitro assays and analytical applications.
  • Potent and selective 5-lipoxygenase inhibitor
  • High purity (99.6%) suitable for analytical use
  • Molecular formula C11H12N2O2S, molecular weight 236.30 g·mol⁻¹
  • Intended for research and in vitro applications
  • Available in small-quantity packs for analytical workflows
Catalog No. 50-002-58928 Supplier Medchemexpress LLC Supplier No. HY1416410MG
Zileuton is a purified small-molecule inhibitor of 5-lipoxygenase used as a research reagent to study leukotriene biology, inflammation, and asthma-related pathways. It is supplied as an analytical standard for in vitro assays and analytical applications.
  • Potent and selective 5-lipoxygenase inhibitor
  • High purity (99.6%) suitable for analytical use
  • Molecular formula C11H12N2O2S, molecular weight 236.30 g·mol⁻¹
  • Intended for research and in vitro applications
  • Available in small-quantity packs for analytical workflows