Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zileuton | 111406-87-2 | MFCD00866097 | 99.61% | C11H12N2O2S | 50 MG
SDP

Catalog No. 5000353338
Encompass_Preferred

Zileuton is a potent and selective inhibitor of 5-lipoxygenase, commonly used in research for its antiasthmatic properties. This chemical appears as a white to off-white solid.

  • Potent and selective inhibitor of 5-lipoxygenase
  • Exhibits antiasthmatic properties
  • For research use only
  • Solid appearance
  • Color: white to off-white
  • Molecular weight: 236.30
  • Melting/freezing point: 157-158°C
  • Relative density: 1.401 g/cm³

Catalog No. 50-003-53338 Supplier Medchemexpress LLC Supplier No. HY1416450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zileuton is a potent and selective inhibitor of 5-lipoxygenase, commonly used in research for its antiasthmatic properties. This chemical appears as a white to off-white solid.

  • Potent and selective inhibitor of 5-lipoxygenase
  • Exhibits antiasthmatic properties
  • For research use only
  • Solid appearance
  • Color: white to off-white
  • Molecular weight: 236.30
  • Melting/freezing point: 157-158°C
  • Relative density: 1.401 g/cm³

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.