Learn More
Selleck Chemical LLC Zileuton S1443-10mM/1mL
SDP
Catalog No. 503110067
Encompass_Preferred
Zileuton is an orally active inhibitor of 5-lipoxygenase and thus inhibits leukotrienes (LTB4 LTC4 LTD4 and LTE4) formation used to decrease the symptoms of asthma Zileuton induces apoptosis while inhibits ferroptosis
Catalog No. 50-311-0067 Supplier Selleck Chemical LLC Supplier No. S144310MM/1ML
Zileuton is an orally active inhibitor of 5-lipoxygenase and thus inhibits leukotrienes (LTB4 LTC4 LTD4 and LTE4) formation used to decrease the symptoms of asthma Zileuton induces apoptosis while inhibits ferroptosis