Learn More
Cayman Chemical ZIleuton-d4 500ug
SDP
Catalog No. 502936954
Encompass_Preferred
An internal standard for the quantification of zileuton by GC- or LC-MS
Catalog No. 50-293-6954 Supplier Cayman Chemical Supplier No. 26761500 UG
An internal standard for the quantification of zileuton by GC- or LC-MS