Learn More
Medchemexpress LLC Zileuton sodium | 118569-21-4 | 98.0% | C11H11N2NaO2S | 100 MG
SDP
Catalog No. 5000353458
Encompass_Preferred
Zileuton sodium is a potent and selective inhibitor of 5-lipoxygenase, exhibiting inflammatory activities.
  • Potent and selective inhibitor of 5-lipoxygenase.
  • Exhibits inflammatory activities.
  • Reduces IL-2 levels by inhibiting 5-lipoxygenase.
  • Decreases NF-κB expression and apoptotic index in I/R rats.
  • Inhibits polyp formation in APCΔ468 colon and small intestine in rats, and increases apoptosis rates in polyps.
Catalog No. 50-003-53458 Supplier Medchemexpress LLC Supplier No. HY14164A100MG
Zileuton sodium is a potent and selective inhibitor of 5-lipoxygenase, exhibiting inflammatory activities.
  • Potent and selective inhibitor of 5-lipoxygenase.
  • Exhibits inflammatory activities.
  • Reduces IL-2 levels by inhibiting 5-lipoxygenase.
  • Decreases NF-κB expression and apoptotic index in I/R rats.
  • Inhibits polyp formation in APCΔ468 colon and small intestine in rats, and increases apoptosis rates in polyps.