Learn More
Medchemexpress LLC Zilganersen sodium | 2305355-64-8 | 91.43% | 7060.8 (free acid) | 1 MG
SDP
Catalog No. 5000396194
Encompass_Preferred
Zilganersen (ION373) sodium is a gapmer antisense oligonucleotide designed to target and inhibit glial fibrillary acidic protein (GFAP) synthesis. It aims to reduce the excess GFAP produced by disease-causing variants in the GFAP gene. This product is suitable for research related to Alexander disease.
  • Targets glial fibrillary acidic protein (GFAP)
  • Reduces excess GFAP from disease-causing variants
  • Inhibits GFAP synthesis
  • Applicable for Alexander disease research
Catalog No. 50-003-96194 Supplier Medchemexpress LLC Supplier No. HYP3392A1MG
Zilganersen (ION373) sodium is a gapmer antisense oligonucleotide designed to target and inhibit glial fibrillary acidic protein (GFAP) synthesis. It aims to reduce the excess GFAP produced by disease-causing variants in the GFAP gene. This product is suitable for research related to Alexander disease.
  • Targets glial fibrillary acidic protein (GFAP)
  • Reduces excess GFAP from disease-causing variants
  • Inhibits GFAP synthesis
  • Applicable for Alexander disease research