Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zilovertamab vedotin | 98.6% | 1 MG
SDP

Catalog No. 5000202766
Encompass_Preferred

Zilovertamab vedotin is an antibody-drug conjugate for preclinical cancer research that combines a humanized anti-ROR1 monoclonal antibody with the cytotoxic payload monomethyl vedotin. It is supplied as a lyophilized powder for experimental use.

  • targets receptor tyrosine kinase-like orphan receptor 1 (ROR1)
  • contains monomethyl vedotin cytotoxic payload
  • suitable for in vitro and in vivo research
  • high reported purity (98.61%)
  • documentation available: certificate of analysis and safety data sheet

Catalog No. 50-002-02766 Supplier Medchemexpress LLC Supplier No. HYP999561MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zilovertamab vedotin is an antibody-drug conjugate for preclinical cancer research that combines a humanized anti-ROR1 monoclonal antibody with the cytotoxic payload monomethyl vedotin. It is supplied as a lyophilized powder for experimental use.

  • targets receptor tyrosine kinase-like orphan receptor 1 (ROR1)
  • contains monomethyl vedotin cytotoxic payload
  • suitable for in vitro and in vivo research
  • high reported purity (98.61%)
  • documentation available: certificate of analysis and safety data sheet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.