Learn More
AdipoGen Zilpaterol hydrochloride
SDP
Catalog No. 502559864
Encompass_Preferred
Chemical. CAS 119520-06-8. Formula C14H19N3O2 . HCl. MW 297.78. Synthetic. Synthetic beta2-adrenoreceptor agonist that binds with the muscle beta-adrenoreceptors, leading to the activation of protein kinase A, an enzyme responsible for changes in protein synthesis and degradation, in particular in skeletal muscles. Due to its partial activity towards beta1-adrenoreceptors, has also the potential to affect cardiac muscle. In consequence, application of zilpaterol leads to preferential induction of fast glycolytic fibre types over slow oxidative fibre types resulting in enhanced growth efficiency and carcass leanness. Used as a veterinary drug in cattle to enhance growth performance in several countries.
Catalog No. 50-255-9864 Supplier AdipoGen Supplier No. CDXZ0025M010
Chemical. CAS 119520-06-8. Formula C14H19N3O2 . HCl. MW 297.78. Synthetic. Synthetic beta2-adrenoreceptor agonist that binds with the muscle beta-adrenoreceptors, leading to the activation of protein kinase A, an enzyme responsible for changes in protein synthesis and degradation, in particular in skeletal muscles. Due to its partial activity towards beta1-adrenoreceptors, has also the potential to affect cardiac muscle. In consequence, application of zilpaterol leads to preferential induction of fast glycolytic fibre types over slow oxidative fibre types resulting in enhanced growth efficiency and carcass leanness. Used as a veterinary drug in cattle to enhance growth performance in several countries.