Learn More
Cayman Chemical ZIlpaterolhydrochlorIde 10mg
SDP
Catalog No. 502971949
Encompass_Preferred
A β-adrenergic receptor agonist that putatively through activation of protein kinase A increases protein synthesis in skeletal muscle fibers, as well as reduces lipogenesis and increases lipolysis in adipose tissues; used as a veterinary food additive to increase lean body weight in commercial beef cattle
Catalog No. 50-297-1949 Supplier Cayman Chemical Supplier No. 1648410 MG
A β-adrenergic receptor agonist that putatively through activation of protein kinase A increases protein synthesis in skeletal muscle fibers, as well as reduces lipogenesis and increases lipolysis in adipose tissues; used as a veterinary food additive to increase lean body weight in commercial beef cattle