Learn More
Selleck Chemical LLC Zilurgisertib fumarate-5MG
SDP
Catalog No. 503134899
Encompass_Preferred
Zilurgisertib fumarate(INCB-000928 fumarate NBU-928 fumarate) is a selective ALK 2 inhibitor It is under development for the treatment of heterotopic ossification and fibrodysplasia ossificans progressiva
Catalog No. 50-313-4899 Supplier Selleck Chemical LLC Supplier No. E15075MG
Zilurgisertib fumarate(INCB-000928 fumarate NBU-928 fumarate) is a selective ALK 2 inhibitor It is under development for the treatment of heterotopic ossification and fibrodysplasia ossificans progressiva