Learn More
Medchemexpress LLC Zimelidine dihydrochloride | 60525-15-7 | 98.08% | 390.15 | 500 MG
SDP
Catalog No. 5000344216
Encompass_Preferred
Zimelidine dihydrochloride is a potent and selective inhibitor of serotonin 5-HT uptake and SERT, acting as an antidepressant.
  • Potent and selective inhibitor of serotonin 5-HT uptake and SERT.
  • Antidepressant.
Catalog No. 50-003-44216 Supplier Medchemexpress LLC Supplier No. HY11002310MG
Zimelidine dihydrochloride is a potent and selective inhibitor of serotonin 5-HT uptake and SERT, acting as an antidepressant.
  • Potent and selective inhibitor of serotonin 5-HT uptake and SERT.
  • Antidepressant.