Learn More
Medchemexpress LLC Zimelidine dihydrochloride | 60525-15-7 | 98.1% | 390.15 | 100 MG
SDP
Catalog No. 5000344822
Encompass_Preferred
Zimelidine dihydrochloride is a potent and selective inhibitor of serotonin 5-HT uptake and SERT. It functions as an antidepressant and is intended for research purposes only. This compound, with a molecular weight of 390.15, appears as a white to off-white solid. It requires specific storage conditions, including -20°C sealed and away from moisture, with particular instructions for solutions.
  • Potent and selective inhibitor of serotonin 5-HT uptake and SERT.
  • Functions as an antidepressant.
  • White to off-white solid appearance.
  • Specific storage at -20°C, sealed and away from moisture.
  • Solution storage at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-003-44822 Supplier Medchemexpress LLC Supplier No. HY110023100MG
Zimelidine dihydrochloride is a potent and selective inhibitor of serotonin 5-HT uptake and SERT. It functions as an antidepressant and is intended for research purposes only. This compound, with a molecular weight of 390.15, appears as a white to off-white solid. It requires specific storage conditions, including -20°C sealed and away from moisture, with particular instructions for solutions.
  • Potent and selective inhibitor of serotonin 5-HT uptake and SERT.
  • Functions as an antidepressant.
  • White to off-white solid appearance.
  • Specific storage at -20°C, sealed and away from moisture.
  • Solution storage at -80°C for 6 months or -20°C for 1 month.