Learn More
Medchemexpress LLC Zimelidine dihydrochloride | 60525-15-7 | 98.3% | 390.15 g/mol | C16H19BrCl2N2 | 5 MG

Supplier: Medchemexpress LLC HY1100235MG
Zimelidine dihydrochloride is a research reagent that functions as a potent and selective serotonin (5-HT) uptake inhibitor targeting the serotonin transporter (SERT). Supplied as the dihydrochloride salt in solid form, it is used in biochemical and pharmacological studies of serotonin transport and receptor function.
- High purity suitable for research applications (98.3%).
- Selective inhibitor of serotonin (5-HT) uptake via SERT.
- Molecular weight 390.15 g/mol; formula C16H19BrCl2N2.
- Supplied as a stable dihydrochloride salt for reproducible dosing.
- Long-term storage recommended at -20°C, sealed and away from moisture.
- Applicable to in vitro and in vivo pharmacology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.