Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine

Catalog No. 38381325MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-381-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-381-325MG Supplier Sigma Aldrich Supplier No. 38381325MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine

Specifications

CAS 107227-89-4
Melting Point 185°C
Molecular Formula C64H80N8O8Zn
Linear Formula C64H80N8O8Zn
Quantity 25 mg
MDL Number MFCD00192579
Synonym 1,4,8,11,15,18,22,25-Octabutoxyphthalocyanine zinc salt
Molecular Weight (g/mol) 1154.76
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.