Learn More
Perkin Elmer US LLC Zinc (Zn) Pure Plus Standard, 1,000 μg/mL, 2% HNO3, 125 mL
Catalog No. 509047928
Encompass_Preferred
Each Single Element Calibration Standard for ICP-MS is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
Catalog No. 50-904-7928 Supplier Perkin Elmer US LLC Supplier No. N9303758
Add to Cart
Add to Cart
Each Single Element Calibration Standard for ICP-MS is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.