Learn More
SPEX CERTIPREP LLC 3P ZINC 2%HNO3 1000PPM 500ML
Catalog No. NC0478471
Encompass_Preferred
(HPS by Zeptometrix) Zn 1000ug/mL in 2% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0478471 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000681500
(HPS by Zeptometrix) Zn 1000ug/mL in 2% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.