Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc 2-Mercaptobenzothiazole 98.0+%, TCI America™
SDP

Catalog No. Z003325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z003325G 25 g
Z0033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z003325G Supplier TCI America Supplier No. Z003325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 155-04-4
Molecular Formula C7H4NS2Zn+
Molecular Weight (g/mol) 231.616
MDL Number MFCD00072234
InChI Key JLKZUNOWPPEBPX-UHFFFAOYSA-M
Synonym 2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate
PubChem CID 25200224
IUPAC Name zinc;1,3-benzothiazole-2-thiolate
SMILES C1=CC=C2C(=C1)N=C(S2)[S-].[Zn+2]

Specifications

Melting Point 325°C
Color White-Yellow
Quantity 25 g
Formula Weight 397.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc 2-Mercaptobenzothiazole
EINECSNumber (5)-3418
RTECSNumber DL7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.