Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc, 99.8% min., ACS, MilliporeSigma™

Catalog No. MZX00107 Shop All MilliporeSigma Products
Change view
Click to view available options
12kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MZX00107 12kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MZX00107 Supplier MilliporeSigma Supplier No. ZX00107

missing translation for 'validMainframeMsg'

Secondary Reference Material For Complexometry

TRUSTED_SUSTAINABILITY

Specifications

Color Silver
CAS 7440-66-6
Density 7.13g/cm3 (20°C)
Quantity 12 kg
Form Granular
Assay Percent Range 99.8% min.
Molecular Formula Zn
Linear Formula Zn
MDL Number MFCD00011291
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES [Zn]
IUPAC Name zinc
Molecular Weight (g/mol) 65.38
PubChem CID 23994
ChEBI CHEBI:30185
Formula Weight 65.39
Grade SRM Certipure
Chemical Name or Material Zinc, Traceable to NIST
Show More Show Less
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.