Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Acetate, 2 N APHA for Sulfide, Reagents

Catalog No. 01333354
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-354 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-354 Supplier Reagents Holdings Llc Supplier No. CZ111500120A
Add to Cart
Add to Cart

Zinc Acetate, 2 N APHA for Sulfide, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 5970-45-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Zinc(II) Acetate
IUPAC Name water
SMILES O

Specifications

Boiling Point 102°C
Color Colorless
pH 6 to 8
Physical Form Liquid
Quantity 120 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1044
Chemical Name or Material Zinc Acetate, 2 N APHA for Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.