Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc acetate | 557-34-6 | 99.1% | 1 ML
SDP

Catalog No. 5000412654
Encompass_Preferred

Zinc acetate is a biological molecule provided as a 10 mM solution in DMSO. This high-purity product is suitable for various laboratory applications.

  • Provided as a 10 mM solution in DMSO.
  • Contains 1 mL of solution.
  • Purity of 99.05%.
  • Data Sheet, COA, and SDS available.
  • Handling instructions provided.
  • Solid form storage: room temperature for 3 years.
  • Solution form storage: -80°C for 2 years, or -20°C for 1 year.

Catalog No. 50-004-12654 Supplier Medchemexpress LLC Supplier No. HYB181710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc acetate is a biological molecule provided as a 10 mM solution in DMSO. This high-purity product is suitable for various laboratory applications.

  • Provided as a 10 mM solution in DMSO.
  • Contains 1 mL of solution.
  • Purity of 99.05%.
  • Data Sheet, COA, and SDS available.
  • Handling instructions provided.
  • Solid form storage: room temperature for 3 years.
  • Solution form storage: -80°C for 2 years, or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.