Learn More
Medchemexpress LLC Zinc acetate | 557-34-6 | 99.99% | 10 G
SDP
Catalog No. 5000412799
Encompass_Preferred
Zinc acetate, 99.99% trace metals basis, also known as Zinc(II) acetate, is a chemical compound primarily used as an alternative staining agent in SDS electrophoresis. It is also recognized for its capability to influence the permeability of biological membranes.
  • High purity of 99.99%
  • Solid appearance
  • White to off-white color
Catalog No. 50-004-12799 Supplier Medchemexpress LLC Supplier No. HYB1817A10G
Zinc acetate, 99.99% trace metals basis, also known as Zinc(II) acetate, is a chemical compound primarily used as an alternative staining agent in SDS electrophoresis. It is also recognized for its capability to influence the permeability of biological membranes.
  • High purity of 99.99%
  • Solid appearance
  • White to off-white color