Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc acetate | 557-34-6 | MFCD00012454 | 99.1% | 100 G
SDP

Catalog No. 5000413053
Encompass_Preferred

Zinc acetate is a biological molecule that appears as a white to off-white solid. It is soluble in DMSO at 50 mg/mL (272.52 mM) with sonication. This product is for research use only and not sold to patients. It has a molecular weight of 183.47 and a chemical formula of Zn(CH3CO2)2.

  • Research use only
  • White to off-white solid appearance
  • Soluble in DMSO
  • Molecular weight of 183.47
  • Chemical formula: Zn(CH3CO2)2

Catalog No. 50-004-13053 Supplier Medchemexpress LLC Supplier No. HYB1817100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc acetate is a biological molecule that appears as a white to off-white solid. It is soluble in DMSO at 50 mg/mL (272.52 mM) with sonication. This product is for research use only and not sold to patients. It has a molecular weight of 183.47 and a chemical formula of Zn(CH3CO2)2.

  • Research use only
  • White to off-white solid appearance
  • Soluble in DMSO
  • Molecular weight of 183.47
  • Chemical formula: Zn(CH3CO2)2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.