Learn More
Medchemexpress LLC Zinc acetate, 99.99% trace metals basis | 557-34-6 | MFCD00012454 | 99.99% | 1 G
SDP
Catalog No. 5000412710
Encompass_Preferred
Zinc acetate, 99.99% trace metals basis, also referred to as Zinc(II) acetate, 99.99% trace metals basis, is a high-purity compound. It is utilized as an alternative staining agent in SDS electrophoresis and can influence the permeability of biological membranes. This product is strictly for research use.
  • Alternative staining agent in SDS electrophoresis
  • Can affect the permeability of biological membranes
  • High purity (99.99% trace metals basis)
Catalog No. 50-004-12710 Supplier Medchemexpress LLC Supplier No. HYB1817A1G
Zinc acetate, 99.99% trace metals basis, also referred to as Zinc(II) acetate, 99.99% trace metals basis, is a high-purity compound. It is utilized as an alternative staining agent in SDS electrophoresis and can influence the permeability of biological membranes. This product is strictly for research use.
  • Alternative staining agent in SDS electrophoresis
  • Can affect the permeability of biological membranes
  • High purity (99.99% trace metals basis)