Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc acetate, anhydrous, 99.9+%

Catalog No. AAH3373409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3373409 10 g
AAH3373418 50 g
AAH3373430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3373409 Supplier Thermo Scientific Chemicals Supplier No. H3373409
Only null left
Add to Cart

CAS: 557-34-6 | C4H6O4Zn | 183.468 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

Solubility
Soluble in water, alcohol, dilute mineral acids and alkalies.

Notes
Incompatible with zinc salts, alkalies and their carbonates, oxalates, phosphates, and sulfides. Hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 557-34-6
Molecular Formula C4H6O4Zn
Molecular Weight (g/mol) 183.468
MDL Number MFCD00012454
InChI Key DJWUNCQRNNEAKC-UHFFFAOYSA-L
Synonym zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
PubChem CID 11192
ChEBI CHEBI:62984
IUPAC Name zinc;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Zn+2]

Specifications

Melting Point 200°C (decomposition)
Linear Formula Zn(OOCCH3)2
Quantity 10 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,dilute mineral acids and alkalies.
Formula Weight 183.47
Percent Purity ≥99.9%
Chemical Name or Material Zinc acetate, anhydrous
Hazard Category H302-H318
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 209-170-2
RTECSNumber AK1500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.