Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Acetate, Crystal, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607815 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-815 12 kg Poly Bottle
18-607-814 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-815 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ZI20012KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZI200, 5970-45-6, C4H6O4Zn·2H2O

Spectrum™ Chemical Zinc Acetate, Crystal, USP is used in anti-itch oitments and for treating zinc deficiencies. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.50
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
IUPAC Name zinc(2+) diacetate dihydrate
SMILES O.O.[Zn++].CC([O-])=O.CC([O-])=O

Specifications

pH 6.0 to 8.0
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP
Packaging Poly Bottle
Chemical Name or Material Zinc Acetate, Crystal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.