Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc acetate dihydrate, 97+%

Catalog No. AAA1290936
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1290936 500 g
AAA1290922 100 g
AAA12909A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1290936 Supplier Thermo Scientific Chemicals Supplier No. A1290936
Add to Cart

CAS: 5970-45-6 | C4H10O6Zn | 219.498 g/mol

Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and alcohol.

Notes
Incompatible with zinc salts, alkalies-their carbonates, oxalates, phosphates and sulfides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate

Specifications

Melting Point 237°C
Density 1.735
pH 6 to 7
Odor Weak, Characteristic
Quantity 500 g
UN Number UN3077
Beilstein 3732513
Merck Index 14,10128
Solubility Information Soluble in water and alcohol.
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
Molecular Weight (g/mol) 219.498
Formula Weight 219.50 (183.47 Anhydrous)
Percent Purity ≥97%
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 209-170-2
RTECSNumber ZG8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.