Learn More
Medchemexpress LLC Zinc acetate, trace metals basis | 557-34-6 | MFCD00012454 | 99.99% | 5 G
SDP
Catalog No. 5000411134
Encompass_Preferred
Zinc acetate, 99.99% trace metals basis, is a chemical compound suitable for various research applications. It can be utilized as an alternative staining agent in SDS electrophoresis and is known to affect the permeability of biological membranes.
  • Can be used as an alternative staining agent in SDS electrophoresis.
  • Affects the permeability of biological membranes.
  • For research use only.
Catalog No. 50-004-11134 Supplier Medchemexpress LLC Supplier No. HYB1817A5G
Zinc acetate, 99.99% trace metals basis, is a chemical compound suitable for various research applications. It can be utilized as an alternative staining agent in SDS electrophoresis and is known to affect the permeability of biological membranes.
  • Can be used as an alternative staining agent in SDS electrophoresis.
  • Affects the permeability of biological membranes.
  • For research use only.