Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc arsenide, 99%, Thermo Scientific™

Catalog No. AAA1762314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1762314 25 g
AAA1762322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1762314 Supplier Thermo Scientific Chemicals Supplier No. A1762314

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12006-40-5
Molecular Formula As2Zn3
Molecular Weight (g/mol) 345.98
MDL Number MFCD00049629
InChI Key RHKSESDHCKYTHI-UHFFFAOYSA-N
Synonym zinc arsenide, trizincarsa-1,4-diene, zinc arsenide, pieces trace metals basis
PubChem CID 25147458
IUPAC Name bis(zincio-λ²-arsanyl) zinc
SMILES [Zn].[Zn]=[As].[Zn]=[As]

Specifications

Melting Point 1015°C
Quantity 25 g
UN Number UN1557
Formula Weight 345.95
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 5.528
Chemical Name or Material Zinc arsenide
Hazard Category H301+H331-H350
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
EINECSNumber 234-486-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.