Learn More
Strem, An Ascensus Company CAS# 7699-45-8. 250g. Zinc bromide (99.9%-Zn). MFCD00011294
SDP
Supplier:  Strem, An Ascensus Company 303032250G
Encompass
CAS# 7699-45-8. 250g. Zinc bromide (99.9%-Zn). MFCD00011294. Molecular Weight: 225.19. Molecular Formula: ZnBr2. Color/form: white pwdr. Strem# 30-3032. http://www.strem.com/catalog/v/30-3032/
Catalog No. 50-901-11387