Learn More
Fisherbrand™ Zinc-Buffered Formalin Prefilled Click Lock Containers
SDP
Catalog No. 23101006 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
23-101-006 20 mL
23-101-007 40 mL
23-101-008 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-101-006
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ready-to-use Buffered Zinc Formalin in leak-resistant, locked containers arranged in convenient cardboard packs
  • Ready-to-use Buffered Zinc Formalin in leak-resistant, locked containers arranged in convenient cardboard packs
  • Recommended for use in IHC, as BFZ preserves more immunoreactivity than standard NBF
  • 20 mL vial contains 10 mL of Buffered Zinc Formalin
Specifications
Product Type Pre-filled Zinc Formalin Container
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 20 mL
Color Clear
No. per Case 96