Learn More
TA Instruments Zinc Calibration Material Not Certified, Standard Zinc, Temperature Calibration, DSC Cells, TMA Instruments, Environmental Systems, Non Certified
Catalog No. 5000580812 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Zinc Calibration Material Not Certified is standard zinc material used for temperature calibration of DSC cells, TMA instruments, and other environmental systems, providing a non-certified reference.
Catalog No. 50-005-80812 Supplier TA Instruments Supplier No. 900907.901
Waters TA Instruments Zinc Calibration Material Not Certified is standard zinc material used for temperature calibration of DSC cells, TMA instruments, and other environmental systems, providing a non-certified reference.