Learn More
Medchemexpress LLC Zinc carbonate | 3486-35-9 | 57.5% | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYB1818
Encompass_Preferred
Zinc monocarbonate is a biological molecule intended for research use only. It is presented as a white to off-white solid, stable under recommended storage conditions, and suitable for various laboratory applications and substance manufacturing.
  • White to off-white solid
  • Stable under recommended storage conditions
  • Soluble at 20 mg/mL in 1 M HCl (with ultrasonic, warming, and heating to 60°C)
  • For research use only
  • Used in laboratory chemicals and manufacturing of substances
Catalog No. 50-004-14323