Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc carbonate | 3486-35-9 | 57.5% | 500 G
SDP

Supplier:  Medchemexpress LLC HYB1818500G

Encompass_Preferred

Zinc monocarbonate is a biological molecule for research use only. It is not intended for patient use.

  • Store at room temperature
  • Keep dry and cool
  • In solvent: -80°C for 1 year, -20°C for 6 months
  • Appearance: solid
  • Color: white to off-white

Catalog No. 50-004-13006


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.