Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Carbonate, MP Biomedicals

Catalog No. ICN10473790 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10473790 500 g
ICN10473780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10473790 Supplier MP Biomedicals, Inc Supplier No. 0210473790
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3486-35-9
Molecular Formula CO3Zn
Molecular Weight (g/mol) 125.388
InChI Key FMRLDPWIRHBCCC-UHFFFAOYSA-L
Synonym zinc carbonate, zincspar, carbonic acid, zinc salt 1:1, zinc monocarbonate, natural smithsonite, zinc carbonate 1:1, unii-k8290ptq4f, hydrozincite?, zinc carbonic acid, zinc 2+ ion carbonate
PubChem CID 19005
IUPAC Name zinc;carbonate
SMILES C(=O)([O-])[O-].[Zn+2]

Specifications

Quantity 500 g
Formula Weight 125.4
Chemical Name or Material Zinc Carbonate
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.