Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Carbonate, Powder, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02004590 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-590 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-590 Supplier Avantor Supplier No. 431201

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 3486-35-9
Molecular Formula CO3Zn
Molecular Weight (g/mol) 125.388
InChI Key FMRLDPWIRHBCCC-UHFFFAOYSA-L
Synonym zinc carbonate, zincspar, carbonic acid, zinc salt 1:1, zinc monocarbonate, natural smithsonite, zinc carbonate 1:1, unii-k8290ptq4f, hydrozincite?, zinc carbonic acid, zinc 2+ ion carbonate
PubChem CID 19005
IUPAC Name zinc;carbonate
SMILES C(=O)([O-])[O-].[Zn+2]

Specifications

Physical Form Powder
Quantity 500 g
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.40kg
Chemical Name or Material Zinc Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.