Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, 0.1 M Solution, Spectrum™ Chemical
SDP

Catalog No. 18607834 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-834 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-834 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z1451LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Z-145, 7646-85-7

Spectrum™ Chemical Zinc Chloride, 0.1 M Solution is a colorless liquid that is mildly corrosive to metals and highly soluble in water. In the laboratory for chemical synthesis, it is used for its properties as a middle strength Lewis acid. It is also used by forensic laboratories for enhanced finger print detection. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 7646-85-7 , 7732-18-5 , 7647-01-0
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
IUPAC Name zinc(2+) dichloride
SMILES [Cl-].[Cl-].[Zn++]

Specifications

CAS Max % 1 to 3%, 80 to 100%, 0.1 - 1
Percent Purity 0.1 M ±0.001 M
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L
Physical Form Chemical Solution
Chemical Name or Material Zinc Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.