Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, 0.100 Molar Standardized, Reagents

Catalog No. 01333350
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-350 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-350 Supplier Reagents Holdings Llc Supplier No. CZ1290004A
Add to Cart
Add to Cart

Zinc Chloride, 0.100 Molar Standardized NIST Traceable, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Zinc Chloride, 0.100 Molar Standardized NIST Traceable
Boiling Point 100°C
CAS 7732-18-5, 7646-85-7
Color Colorless
pH 3.9 to 4.1
Physical Form Liquid
Quantity 4 L
Synonym Zinc Dichloride
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0124
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.