Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc Chloride
SCI_ED

Catalog No. S25635A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25635A 100 g Reagent Grade
S25635 500 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25635A

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Color White
Physical Form Solid
Quantity 100 g
Assay Percent Range >95%
Linear Formula ZnCl2
Formula Weight 136.29
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Zinc Chloride
Safety handling Check Alert storage code Red
ShelfLife 1 year
DOTInformation DOT Class 8, III, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.