Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride, MilliporeSigma™

Catalog No. M1088161000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1088161000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1088161000 Supplier MilliporeSigma Supplier No. 1.08816.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For analysis EMSURE™ ACS, ISO, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Boiling Point 732°C
Quantity 1 kg
UN Number UN2331
Solubility Information Soluble in water: 4320g/L at 25°C
Formula Weight 136.3
Odor Odorless
Packaging Plastic Bottle
Vapor Pressure 1.33hPa at 428°C
Density 2.91g/cm3 at 25°C
Chemical Name or Material Zinc chloride
Hazard Category H302: Harmful if swallowed.
Hazard Statement H314: Causes severe skin burns and eye damage.
Precautionary Statement H410: Very toxic to aquatic life with long lasting effects.
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-592-0
RTECSNumber ZH1400000
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.