Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Granular, USP, 97-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607844 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-844 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-844 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ZI10550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ZI105, 7646-85-7, ZnCl2

Spectrum™ Chemical Zinc Chloride, Granular, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

CAS 7646-85-7
CAS Max % 1
Molecular Formula Cl2Zn
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Molecular Weight (g/mol) 136.28
Percent Purity 97 to 100.5%
Grade USP
Packaging Fiber Drum
Quantity 50 kg
Chemical Name or Material Zinc Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.