Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride, MP Biomedicals™

Catalog No. MP219145190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219145190 500 g
MP219145191 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219145190 Supplier MP Biomedicals, Inc Supplier No. 0219145190
Add to Cart
Add to Cart

Zinc chloride ACS reagent grade, Hard, White Crystalline Powder

  • It is a widely used reagent in various industrial processes.
  • These include textile finishing, the etching of metals, the manufacture of parchment paper, and the preparation of galvanizing, soldering, and tinning fluxes.
  • It is also used in organic chemical synthesis, such as in the formation of amines from azides.
  • It is used in biochemistry and molecular biology research as a source of zinc ion.
  • A study has been published on the zinc mediated inhibition of cAMP production in N18TG2 neuroblastoma cells.
  • The use of zinc chloride in crystal studies of cytochrome c oxidase and of cytochrome c oxidoreductase has been described.
  • Zinc chloride has been utilized to investigate the relationship of glutathione and glutathione synthesis rates to cellular toxicity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym Butter of zinc, Chlorure de zinc, Chlorure de zinc anhydre, Cloruro de cinc
PubChem CID 5727
ChEBI CHEBI:49976

Specifications

Boiling Point 732°C (literature)
Melting Point 290°C (literature)
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range >97.0%
UN Number UN2331
Solubility Information This product is soluble in 0.029 M HCl (97mg/mL), yielding a clear to slightly hazy, colorless solution. It is also soluble in alcohol,glycerol, and acetone and water.
Vapor Pressure 1mmHg (428°C) (Lit)
Density 2.907g/mL(Lit.)
Assay ACS reagentgrade,
Show More Show Less
RTECSNumber ZH1400000
storageNote1 Store at Room Temperature (15-30°C). Store Desiccated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.