Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, puriss., Solstice

Catalog No. 6001783 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-83 500 g
60-044-857 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-017-83 Supplier Solstice Supplier No. 14422500G

Puriss., meets analytical specification of BP, Ph. Eur., USP, 98-100.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 500 g
Linear Formula ZnCl2
UN Number 2331
Formula Weight 136.3g/mol
Percent Purity 98 to 100.5%
Grade puriss.
Vapor Pressure 1mmHg (428°C)
Chemical Name or Material Zinc Chloride, Meets analytical specification of Ph. Eur.BP, USP
missing translation for 'healthHazard2' P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'healthHazard3' H302-H314-H410
EINECSNumber 231-592-0
RTECSNumber ZH1400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.