Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001779 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-79 25 kg
60-017-77 1 kg
60-017-76 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-017-79 Supplier Solstice Supplier No. 3165025KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 25 kg
Linear Formula ZnCl2
UN Number 2331
Formula Weight 136.3g/mol
Percent Purity ≥98%
Grade ACS/puriss. p.a.
Vapor Pressure 1mmHg (428°C)
Chemical Name or Material Zinc Chloride, Reag. Ph. Eur. reag. ISO
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302-H314-H410
EINECSNumber 231-592-0
RTECSNumber ZH1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.