Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Citrate, Dihydrate, 97%, Spectrum™ Chemical
SDP

Catalog No. 18614736 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-736 25 g Poly Bottle
18-614-737 100 g Poly Bottle
18-614-739 500 g Poly Bottle
18-614-741 2.5 kg Poly Pail
18-614-738 12 kg Poly Pail
18-614-740 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-614-736 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z105225GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5990-32-9
Molecular Formula C12H14O16Zn3
Molecular Weight (g/mol) 610.37
InChI Key OXAGUGIXGVHDGD-UHFFFAOYSA-H
IUPAC Name trizinc(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate) dihydrate
SMILES O.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Quantity 25 g
Solubility Information To Pass Test
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.