Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Citrate, Dihydrate, 97%, Spectrum™ Chemical
SDP

Catalog No. 18614741 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-741 2.5 kg Poly Bottle
18-614-736 25 g Poly Bottle
18-614-737 100 g Poly Bottle
18-614-739 500 g Poly Bottle
18-614-738 12 kg Poly Bottle
18-614-740 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-614-741 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z10522.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5990-32-9
Molecular Formula C12H14O16Zn3
Molecular Weight (g/mol) 610.37
InChI Key OXAGUGIXGVHDGD-UHFFFAOYSA-H
IUPAC Name trizinc(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate) dihydrate
SMILES O.O.[Zn++].[Zn++].[Zn++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Solubility Information To Pass Test
Percent Purity 97%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Zinc Citrate, Dihydrate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.