Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Citrate, MP Biomedicals

Catalog No. ICN219811 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219811 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219811 Supplier MP Biomedicals, Inc Supplier No. 0521981180

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5990-32-9
Molecular Formula C12H14O16Zn3
Molecular Weight (g/mol) 610.368
InChI Key OXAGUGIXGVHDGD-UHFFFAOYSA-H
Synonym zinc citrate dihydrate, unii-021ibv57il, acmc-20aj8r, citric acid zinc salt dihydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt 2:3 , dihydrate, trizinc 2+ dicitrate dihydrate, zinc citrate, dihydrate, reagent, trizinc, ion zn2+ dicitrate dihydrate, zinc 2-hydroxypropane-1,2,3-tricarboxylate-water 3/2/2, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt, hydrate 2:3:2
PubChem CID 16213234
IUPAC Name trizinc;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Zn+2].[Zn+2].[Zn+2]

Specifications

Quantity 100 g
Formula Weight 610.35
Chemical Name or Material Zinc Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.