Learn More
Bioworld Zinc-Coated Microplates, 5 Plate(s)
Catalog No. 50197553
Encompass
Zinc coated microplates use a chelatingsurface charged with divalent cobalt (Zn 2+) to enable specific, fast and gentle purification of histidine-tagged fusion proteins from a variety of sample types.
Catalog No. 50-197-553 Supplier Bioworld Supplier No. 201400221
Zinc coated microplates use a chelatingsurface charged with divalent cobalt (Zn 2+) to enable specific, fast and gentle purification of histidine-tagged fusion proteins from a variety of sample types.